Assay Design
Gene Transcripts
- Gene Symbol
- Otub2
- Entrez Gene ID
-
68149
- Gene Name
- OTU domain, ubiquitin aldehyde binding 2
- Gene Aliases
-
2010015L18Rik, 4930586I02Rik, AI413508, AW557219, OTB2, OTU2
- Chromosome Location
-
Chr.12: 103376894 - 103406351 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK008258.2 |
- |
1 - 2 |
83 |
84 |
View Full Product Details