Assay Design
Gene Transcripts
- Gene Symbol
- Tmed9
- Entrez Gene ID
-
67511
- Gene Name
- transmembrane emp24 protein transport domain containing 9
- Gene Aliases
-
2400003B06Rik
- Chromosome Location
-
Chr.13: 55593135 - 55597697 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK044120.1 |
- |
1 - 2 |
84 |
70 |
View Full Product Details