Assay Design



Gene Transcripts

Gene Symbol
Optn
Entrez Gene ID
71648
Gene Name
optineurin
Gene Aliases
4930441O07Rik, FIP2, HYPL, NRP
Chromosome Location
Chr.2: 5020520 - 5064112 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_181848.4 NP_862896.1 3 - 4 410 88
GenBank mRNA AY071834.1 - 3 - 4 517 88
BC061185.1 - 3 - 4 410 88

View Full Product Details