Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001290576.1 | NP_001277505.1 | 4 - 5 | 830 | 97 |
NM_016768.2 | NP_058048.1 | 4 - 5 | 830 | 97 | |
GenBank mRNA | AF020199.1 | - | 4 - 5 | 821 | 97 |
AF020200.1 | - | 4 - 5 | 823 | 97 | |
AK144363.1 | - | 4 - 5 | 747 | 97 | |
BC057879.1 | - | 4 - 5 | 721 | 97 |