Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001290576.1 | NP_001277505.1 | 7 - 8 | 1229 | 82 |
NM_016768.2 | NP_058048.1 | 8 - 9 | 1342 | 82 | |
GenBank mRNA | AF020199.1 | - | 8 - 9 | 1333 | 82 |
AF020200.1 | - | 7 - 8 | 1222 | 82 | |
AK144363.1 | - | 7 - 8 | 1146 | 82 | |
BC057879.1 | - | 8 - 9 | 1233 | 82 |