Assay Design



Gene Transcripts

Gene Symbol
Tmem236
Entrez Gene ID
625286
Gene Name
transmembrane protein 236
Gene Aliases
2010003H20Rik, Fam23a, Gm613
Chromosome Location
Chr.2: 14174524 - 14221993 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081310.2 NP_001074779.1 2 - 3 384 69
GenBank mRNA AK144489.1 - 2 - 3 385 69
BY733764.1 - 2 - 3 385 69

View Full Product Details