Assay Design



Gene Transcripts

Gene Symbol
Vav2
Entrez Gene ID
22325
Gene Name
vav 2 oncogene
Gene Aliases
2810040F13Rik, AI847175
Chromosome Location
Chr.2: 27262104 - 27427608 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009500.1 NP_033526.1 10 - 11 1375 86
GenBank mRNA BC053060.1 - 10 - 11 1352 86
U37017.1 - 10 - 11 1375 86

View Full Product Details