Assay Design
Gene Transcripts
- Gene Symbol
- Gabrg3
- Entrez Gene ID
-
14407
- Gene Name
- gamma-aminobutyric acid (GABA) A receptor, subunit gamma 3
- Gene Aliases
-
B230362M20Rik, Gabrg-3
- Chromosome Location
-
Chr.7: 56724242 - 57386871 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
X59300.1 |
- |
8 - 9 |
1090 |
85 |
View Full Product Details