Assay Design



Gene Transcripts

Gene Symbol
Fars2
Entrez Gene ID
69955
Gene Name
phenylalanine-tRNA synthetase 2 (mitochondrial)
Gene Aliases
Fars1, pheRS
Chromosome Location
Chr.13: 36117643 - 36537595 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001039189.2 NP_001034278.1 1 - 2 365 68
GenBank mRNA AK020164.1 - 1 - 2 151 68

View Full Product Details