Assay Design



Gene Transcripts

Gene Symbol
Atg13
Entrez Gene ID
51897
Gene Name
autophagy related 13
Gene Aliases
1110053A20Rik, D2Ertd391e, Harbi1
Chromosome Location
Chr.2: 91674612 - 91710598 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_145528.3 NP_663503.1 12 - 13 1398 75
GenBank mRNA AK122339.1 - 8 - 9 3760 75
AK149672.1 - 12 - 13 1357 75
AK154470.1 - 12 - 13 1386 75
AK163962.1 - 12 - 13 1352 75
AK171206.1 - 12 - 13 1384 75
AK171440.1 - 12 - 13 1375 75
BC016669.1 - 12 - 13 1382 75
BC023673.1 - 12 - 13 1348 75
BC023712.1 - 12 - 13 1371 75
BC033419.1 - 12 - 13 1349 75

View Full Product Details