Assay Design



Gene Transcripts

Gene Symbol
Apopt1
Entrez Gene ID
68020
Gene Name
apoptogenic, mitochondrial 1
Gene Aliases
1700081D05Rik, 2810002N01Rik, Apop-1, Apop1
Chromosome Location
Chr.12: 111713269 - 111755055 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001163388.1 NP_001156860.1 4 - 5 503 97

View Full Product Details