Assay Design
Gene Transcripts
- Gene Symbol
- Ankrd9
- Entrez Gene ID
-
74251
- Gene Name
- ankyrin repeat domain 9
- Gene Aliases
-
2500003O20Rik
- Chromosome Location
-
Chr.12: 110975353 - 110979021 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK010867.1 |
- |
3 - 3 |
|
179 |
AK172167.1 |
- |
3 - 4 |
451 |
64 |
View Full Product Details