Assay Design
Gene Transcripts
- Gene Symbol
- Ppig
- Entrez Gene ID
-
228005
- Gene Name
- peptidyl-prolyl isomerase G (cyclophilin G)
- Gene Aliases
-
AU019516, AU022200, B230312B02Rik, CYP, SRCyp
- Chromosome Location
-
Chr.2: 69722617 - 69754059 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK088007.1 |
- |
1 - 2 |
84 |
63 |
BC016126.1 |
- |
1 - 2 |
135 |
63 |
BC048905.1 |
- |
1 - 2 |
133 |
63 |
View Full Product Details