Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001290782.1 | NP_001277711.1 | 14 - 15 | 1588 | 61 |
NM_183220.3 | NP_899043.1 | 14 - 15 | 1643 | 61 | |
GenBank mRNA | AK011867.1 | - | 14 - 15 | 60 | |
BC039569.1 | - | 14 - 15 | 1620 | 61 |