Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_029911.4 | NP_084187.2 | 6 - 7 | 1099 | 62 |
GenBank mRNA | AK031904.1 | - | 6 - 7 | 1030 | 62 |
AK036066.1 | - | 6 - 7 | 1373 | 62 | |
AK082153.1 | - | 6 - 7 | 1100 | 62 | |
BC132487.1 | - | 6 - 7 | 1041 | 62 | |
BC137869.1 | - | 6 - 7 | 1041 | 62 | |
DQ185134.1 | - | 6 - 7 | 996 | 62 |