Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_181416.3 | NP_852081.2 | 4 - 5 | 1118 | 106 |
GenBank mRNA | AK129034.1 | - | - | 1113 | 106 |
AK137111.1 | - | 4 - 5 | 755 | 106 | |
BC050875.1 | - | 4 - 5 | 1072 | 106 | |
BC058105.1 | - | 4 - 5 | 1118 | 106 |