Assay Design



Gene Transcripts

Gene Symbol
Akr1c6
Entrez Gene ID
83702
Gene Name
aldo-keto reductase family 1, member C6
Gene Aliases
3alpha-HSD, Akr1c1, Hsd17b5
Chromosome Location
Chr.13: 4434311 - 4457530 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_030611.3 NP_085114.1 2 - 3 304 76
GenBank mRNA AK149458.1 - 2 - 3 305 76
BC056643.1 - 2 - 3 286 76
D45850.1 - 2 - 3 256 76

View Full Product Details