Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_010067.4 | NP_034197.3 | 5 - 6 | 424 | 149 |
GenBank mRNA | AF012129.1 | - | 5 - 6 | 411 | 149 |
AF045889.1 | - | 5 - 6 | 416 | 149 | |
AK029136.1 | - | 5 - 6 | 425 | 149 | |
AK050403.1 | - | 5 - 6 | 424 | 149 | |
BC087892.1 | - | 5 - 6 | 424 | 149 |