Assay Design



Gene Transcripts

Gene Symbol
Anapc1
Entrez Gene ID
17222
Gene Name
anaphase promoting complex subunit 1
Gene Aliases
2610021O03Rik, AI047775, AI853536, AW547281, Apc1, Mcpr, tsg24
Chromosome Location
Chr.2: 128610083 - 128687410 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_008569.2 NP_032595.2 23 - 24 3009 71
GenBank mRNA AK052404.1 - 23 - 24 2990 71
AK054305.1 - 23 - 24 2991 71
BC139002.1 - 23 - 24 2990 71
BC145492.1 - 23 - 24 2990 71
X80169.1 - 23 - 24 2994 71

View Full Product Details