Assay Design



Gene Transcripts

Gene Symbol
Il1a
Entrez Gene ID
16175
Gene Name
interleukin 1 alpha
Gene Aliases
Il-1a
Chromosome Location
Chr.2: 129299609 - 129310901 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_010554.4 NP_034684.2 4 - 5 378 63
GenBank mRNA AK156449.1 - 4 - 5 379 63
BC003727.1 - 3 - 4 347 63
X01450.1 - 4 - 5 378 63

View Full Product Details