Assay Design



Gene Transcripts

Gene Symbol
Trip11
Entrez Gene ID
109181
Gene Name
thyroid hormone receptor interactor 11
Gene Aliases
2610511G22Rik, 3110031G15Rik, 6030460N08Rik, AI450776, GMAP-210, TRIP230
Chromosome Location
Chr.12: 101834043 - 101913649 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_028446.1 NP_082722.1 10 - 11 1871 117
GenBank mRNA AK081763.1 - 10 - 11 1784 117
AK148294.1 - 10 - 11 1847 117
BC158098.1 - 10 - 11 1967 117

View Full Product Details