Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001136081.2 | NP_001129553.1 | 14 - 15 | 1982 | 80 |
NM_182990.4 | NP_892035.2 | 14 - 15 | 2111 | 80 | |
GenBank mRNA | AK146446.1 | - | 14 - 15 | 2034 | 80 |
AK146585.1 | - | 14 - 15 | 2031 | 80 | |
AK146607.1 | - | 14 - 15 | 2014 | 80 | |
AK146691.1 | - | 14 - 15 | 2013 | 80 | |
AK147005.1 | - | 14 - 15 | 2042 | 80 | |
AK158552.1 | - | 14 - 15 | 1982 | 80 | |
S50213.1 | - | 13 - 14 | 1856 | 80 |