Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_009157.4 | NP_033183.1 | 10 - 11 | 1127 | 84 |
GenBank mRNA | AK036928.1 | - | 10 - 11 | 1131 | 84 |
AK044534.1 | - | 10 - 11 | 1119 | 84 | |
AK044698.1 | - | 10 - 11 | 1119 | 84 | |
AK139777.1 | - | 11 - 12 | 1150 | 84 | |
BC029833.1 | - | 8 - 9 | 878 | 84 | |
U18310.1 | - | 10 - 11 | 1102 | 84 |