Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_172521.1 | NP_766109.1 | 5 - 6 | 1385 | 120 |
GenBank mRNA | AK077049.1 | - | 5 - 6 | 1385 | 120 |
BC125332.1 | - | 5 - 6 | 1365 | 120 | |
BC125334.1 | - | 5 - 6 | 1365 | 120 |