Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001198808.1 | NP_001185737.1 | 3 - 4 | 297 | 69 |
NM_001198809.1 | NP_001185738.1 | 3 - 4 | 376 | 69 | |
NM_177386.5 | NP_796360.2 | 4 - 5 | 420 | 69 | |
GenBank mRNA | AK040600.1 | - | 3 - 4 | 377 | 69 |
AK049505.1 | - | 3 - 4 | 239 | 69 | |
AK052339.1 | - | 3 - 4 | 363 | 69 | |
AK147651.1 | - | 4 - 5 | 275 | 69 | |
AK168444.1 | - | 5 - 6 | 325 | 69 | |
AK220408.1 | - | 1 - 2 | 162 | 69 | |
BB636293.1 | - | 3 - 4 | 377 | 69 | |
BC115510.1 | - | 3 - 4 | 231 | 69 | |
BC115511.1 | - | 3 - 4 | 231 | 69 | |
BC139354.1 | - | 3 - 4 | 208 | 69 | |
BC145610.1 | - | 2 - 3 | 164 | 69 |