Assay Design



Gene Transcripts

Gene Symbol
Wt1
Entrez Gene ID
22431
Gene Name
Wilms tumor 1 homolog
Gene Aliases
D630046I19Rik, Wt-1
Chromosome Location
Chr.2: 105126529 - 105173614 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_144783.2 NP_659032.3 3 - 4 1145 92
GenBank mRNA DQ537939.1 - 3 - 4 482 92
M55512.1 - 3 - 4 1150 92

View Full Product Details