Assay Design



Gene Transcripts

Gene Symbol
Cntrob
Entrez Gene ID
216846
Gene Name
centrobin, centrosomal BRCA2 interacting protein
Gene Aliases
9830165K03Rik, Lip8
Chromosome Location
Chr.11: 69299491 - 69323893 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_172560.3 NP_766148.2 2 - 3 1270 61
GenBank mRNA AK147774.1 - 2 - 3 1156 61
BC058512.1 - 2 - 3 103 61

View Full Product Details