Assay Design



Gene Transcripts

Gene Symbol
Pax6
Entrez Gene ID
18508
Gene Name
paired box 6
Gene Aliases
1500038E17Rik, AEY11, Dey, Gsfaey11, Pax-6, Sey
Chromosome Location
Chr.2: 105667518 - 105697364 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001244200.1 NP_001231129.1 2 - 3 365 56
GenBank mRNA CF725212.1 - 1 - 2 53 56
X63963.1 - 1 - 2 30 56

View Full Product Details