Assay Design
Gene Transcripts
- Gene Symbol
- Ube2s
- Entrez Gene ID
-
77891
- Gene Name
- ubiquitin-conjugating enzyme E2S
- Gene Aliases
-
0910001J09Rik, 6720465F12Rik, AA409170, E2-EPF
- Chromosome Location
-
Chr.7: 4808014 - 4812340 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK020143.1 |
- |
- |
939 |
63 |
View Full Product Details