Assay Design



Gene Transcripts

Gene Symbol
Pax1
Entrez Gene ID
18503
Gene Name
paired box 1
Gene Aliases
Pax-1, hbs, hunchback, un, undulated, wt
Chromosome Location
Chr.2: 147361925 - 147375049 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_008780.2 NP_032806.2 3 - 4 1092 89
GenBank mRNA AK134066.1 - 3 - 4 1061 89
BC140989.1 - 3 - 4 889 89

View Full Product Details