Assay Design
Gene Transcripts
- Gene Symbol
- Neurog1
- Entrez Gene ID
-
18014
- Gene Name
- neurogenin 1
- Gene Aliases
-
AKA, Math4C, Neurod3, bHLHa6, ngn1
- Chromosome Location
-
Chr.13: 56250492 - 56252163 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_010896.2 |
NP_035026.1 |
1 - 1 |
947 |
127 |
GenBank mRNA |
BC062148.1 |
- |
1 - 1 |
947 |
127 |
View Full Product Details