Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025613.3 | NP_079889.2 | 1 - 1 | 1475 | 174 |
GenBank mRNA | AK002559.1 | - | 1 - 1 | 1480 | 174 |
AK005258.1 | - | 1 - 1 | 1445 | 174 | |
AK017543.1 | - | 1 - 1 | 1448 | 174 | |
AK077783.1 | - | 1 - 1 | 1456 | 174 | |
AK078409.1 | - | 1 - 1 | 1421 | 174 | |
AK148555.1 | - | 1 - 1 | 1418 | 174 | |
AK210334.1 | - | 1 - 1 | 292 | 174 | |
AL357452.1 | - | 1 - 1 | 941 | 174 | |
AL357453.1 | - | 1 - 1 | 1441 | 174 | |
DV050753.1 | - | 1 - 1 | 236 | 174 |