Assay Design



Gene Transcripts

Gene Symbol
Crb2
Entrez Gene ID
241324
Gene Name
crumbs family member 2
Gene Aliases
5930402A21, BC043114
Chromosome Location
Chr.2: 37776225 - 37799103 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001163566.1 NP_001157038.1 7 - 8 2029 73
GenBank mRNA BC043114.1 - 6 - 7 1717 73

View Full Product Details