Assay Design



Gene Transcripts

Gene Symbol
Arid1b
Entrez Gene ID
239985
Gene Name
AT rich interactive domain 1B (SWI-like)
Gene Aliases
8030481M12, 9330189K18Rik, AI836955, Ardi1b, B230217J03Rik, mKIAA1235
Chromosome Location
Chr.17: 4992755 - 5347656 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001085355.1 NP_001078824.1 11 - 12 3141 56
GenBank mRNA CK389103.1 - 1 - 2 56

View Full Product Details