Assay Design
Gene Transcripts
- Gene Symbol
- Tmco5b
- Entrez Gene ID
-
75275
- Gene Name
- transmembrane and coiled-coil domains 5B
- Gene Aliases
-
4930563P21Rik
- Chromosome Location
-
Chr.2: 113285095 - 113297190 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_029232.2 |
NP_083508.1 |
4 - 5 |
434 |
77 |
GenBank mRNA |
BC050803.1 |
- |
4 - 5 |
434 |
77 |
View Full Product Details