Assay Design



Gene Transcripts

Gene Symbol
Ciao1
Entrez Gene ID
26371
Gene Name
cytosolic iron-sulfur protein assembly 1
Gene Aliases
AW210570, Wdr39
Chromosome Location
Chr.2: 127240938 - 127247816 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_025296.4 NP_079572.2 3 - 4 598 77
GenBank mRNA AK002314.1 - 3 - 4 543 77
AK028376.1 - 3 - 4 525 77
BC004089.1 - 3 - 4 519 77
BI734652.1 - 3 - 4 598 77

View Full Product Details