Assay Design



Gene Transcripts

Gene Symbol
Ak7
Entrez Gene ID
78801
Gene Name
adenylate kinase 7
Gene Aliases
4930502N02Rik, AK 7
Chromosome Location
Chr.12: 105703859 - 105782447 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_030187.1 NP_084463.1 2 - 3 480 74
GenBank mRNA AK019664.1 - 2 - 3 345 74
AK029555.1 - 2 - 3 481 74

View Full Product Details