Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_033268.4 | NP_150371.4 | 3 - 4 | 591 | 64 |
GenBank mRNA | AF248643.1 | - | 3 - 4 | 359 | 64 |
AK047768.1 | - | 3 - 4 | 592 | 64 | |
AY036877.1 | - | 4 - 5 | 535 | 64 | |
BC089579.1 | - | 3 - 4 | 591 | 64 |