Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001199043.1 | NP_001185972.1 | 1 - 2 | 496 | 91 |
GenBank mRNA | AK004782.2 | - | 1 - 2 | 178 | 91 |
AK029685.1 | - | 1 - 2 | 205 | 91 | |
BC040243.1 | - | 1 - 2 | 144 | 91 |