Assay Design



Gene Transcripts

Gene Symbol
Slc38a11
Entrez Gene ID
320106
Gene Name
solute carrier family 38, member 11
Gene Aliases
9330158F14Rik
Chromosome Location
Chr.2: 65315799 - 65364394 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_177074.2 NP_796048.2 4 - 5 474 121
GenBank mRNA AK043610.1 - 4 - 5 483 121
AK139966.1 - 4 - 5 424 121

View Full Product Details