Assay Design



Gene Transcripts

Gene Symbol
Spag6l
Entrez Gene ID
381350
Gene Name
sperm associated antigen 6 like
Gene Aliases
-
Chromosome Location
Chr.2: 18694011 - 18749746 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001001334.2 NP_001001334.2 10 - 11 1500 110
GenBank mRNA AK132882.1 - 10 - 11 1500 110
BC061194.1 - 10 - 11 1497 110

View Full Product Details