Assay Design



Gene Transcripts

Gene Symbol
Osbpl10
Entrez Gene ID
74486
Gene Name
oxysterol binding protein-like 10
Gene Aliases
4933433D06Rik, C820004B04Rik, OPR-10
Chromosome Location
Chr.9: 114978583 - 115233273 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_148958.2 NP_683761.1 2 - 3 245 57
GenBank mRNA AK017034.1 - 4 - 5 576 57
BC027256.1 - 2 - 3 245 57

View Full Product Details