Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_173033.3 | NP_766621.3 | 9 - 10 | 1385 | 83 |
GenBank mRNA | AK028258.1 | - | 9 - 10 | 1328 | 83 |
AK155456.1 | - | 9 - 10 | 1332 | 83 | |
BC057893.1 | - | 9 - 10 | 1320 | 83 |