Assay Design



Gene Transcripts

Gene Symbol
Igsf21
Entrez Gene ID
230868
Gene Name
immunoglobulin superfamily, member 21
Gene Aliases
BC055811, Gm141
Chromosome Location
Chr.4: 140026852 - 140246811 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_198610.2 NP_941012.1 7 - 8 1544 76
GenBank mRNA AK137439.1 - 7 - 8 1541 76
BC055811.1 - 7 - 8 1517 76

View Full Product Details