Assay Design
Gene Transcripts
- Gene Symbol
- Sh2d5
- Entrez Gene ID
-
230863
- Gene Name
- SH2 domain containing 5
- Gene Aliases
-
BC036961
- Chromosome Location
-
Chr.4: 138250411 - 138260968 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BC036961.1 |
- |
1 - 2 |
55 |
76 |
View Full Product Details