Assay Design



Gene Transcripts

Gene Symbol
Tekt5
Entrez Gene ID
70426
Gene Name
tektin 5
Gene Aliases
3300001K11Rik
Chromosome Location
Chr.16: 10357952 - 10395465 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001099275.1 NP_001092745.1 2 - 3 684 126
NM_001291001.1 NP_001277930.1 2 - 3 701 126
GenBank mRNA AK014352.1 - 2 - 3 553 126
BC115969.1 - 2 - 3 684 126
GQ292766.1 - 2 - 3 701 126

View Full Product Details