Assay Design
Gene Transcripts
- Gene Symbol
- Slx4
- Entrez Gene ID
-
52864
- Gene Name
- SLX4 structure-specific endonuclease subunit homolog (S. cerevisiae)
- Gene Aliases
-
AI256635, AI426760, Btbd12, D16Bwg1016e
- Chromosome Location
-
Chr.16: 3979105 - 4003829 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_177472.5 |
NP_803423.2 |
3 - 4 |
723 |
103 |
GenBank mRNA |
BC065125.1 |
- |
3 - 4 |
728 |
103 |
View Full Product Details