Assay Design



Gene Transcripts

Gene Symbol
Scn11a
Entrez Gene ID
24046
Gene Name
sodium channel, voltage-gated, type XI, alpha
Gene Aliases
NSS2, NaN, NaT, NaV1.9, SNS2
Chromosome Location
Chr.9: 119753730 - 119825456 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_011887.3 NP_036017.3 5 - 6 665 90
GenBank mRNA AB031389.1 - 5 - 6 723 90
AF118044.1 - 5 - 6 665 90

View Full Product Details