Assay Design



Gene Transcripts

Gene Symbol
Rgn
Entrez Gene ID
19733
Gene Name
regucalcin
Gene Aliases
AI265316, GNL, SMP30
Chromosome Location
Chr.X: 20549767 - 20562089 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009060.2 NP_033086.1 2 - 3 234 67
GenBank mRNA AK146465.1 - 2 - 3 266 67
BC012710.1 - 2 - 3 247 67
D86217.1 - 1 - 2 186 67
U28937.1 - 2 - 3 234 67

View Full Product Details