Assay Design



Gene Transcripts

Gene Symbol
Muc13
Entrez Gene ID
17063
Gene Name
mucin 13, epithelial transmembrane
Gene Aliases
114/A10, 14/A10, AI159736, Lrrp, Ly64, NJ-1
Chromosome Location
Chr.16: 33794037 - 33819929 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_010739.2 NP_034869.1 2 - 3 248 75
GenBank mRNA AK132274.1 - 1 - 2 93 75
BC024321.1 - 1 - 2 214 75
J04634.1 - 2 - 3 248 75

View Full Product Details